• USA Home
  • B1507 - tert-Butyl(chloro)dimethylsilane

EMAIL THIS PAGE TO A FRIEND
B1507 Sigma

tert-Butyl(chloro)dimethylsilane

Synonym: tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMSCl

Purchase

Properties

InChI Key   BCNZYOJHNLTNEZ-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2925 8(4.1) / PGII
WGK Germany 
2
RTECS 
VV2000000
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

190500 tert-Butyldimethylsilyl chloride, reagent grade, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?