• USA Home
  • B152 - N6-Benzyl-5′-N-ethylcarboxamidoadenosine

EMAIL THIS PAGE TO A FRIEND
B152 Sigma

N6-Benzyl-5′-N-ethylcarboxamidoadenosine

solid

Synonym: N6-Benzyl-NECA

  • Empirical Formula (Hill Notation) C19H22N6O4

  • Molecular Weight 398.42

  •  MDL number MFCD00274027

  •  PubChem Substance ID 329773189

Purchase

Properties

Related Categories Adenosines/P2 Nucleotide Receptors (Purinergics), Agonists, Cell Biology, Cell Signaling and Neuroscience, Neuroscience,
InChI Key   VVHDYJFDZYHAMR-QCUYGVNKSA-N
form   solid
color   white
solubility   45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: soluble1.28 mg/mL
storage temp.   2-8°C
Gene Information   human ... ADORA3(140)

Description

Biochem/physiol Actions

Selective A3 adenosine receptor agonist.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?