• USA Home
  • B1627 - Brain Extract from bovine brain

EMAIL THIS PAGE TO A FRIEND
B1627 Sigma

Brain Extract from bovine brain

Type III, Folch Fraction III

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycerolipids, Lipids, Mix
type   Type III
storage temp.   −20°C

Description

Other Notes

Contains approx. 80% phosphatidylserine; balance primarily other brain lipids.

Packaging

1 g in ampule

100 mg in ampule

Sealed ampule.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?