EMAIL THIS PAGE TO A FRIEND
B1759 Sigma

sec-Butyl disulfide

~95%

Synonym: Di-sec-butyl disulfide

Purchase

Properties

InChI Key   QTWKINKGAHTPFJ-UHFFFAOYSA-N
assay   ~95%
refractive index   n20/D 1.492(lit.)
density   0.957 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B94004 sec-Butyl disulfide, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?