• USA Home
  • B1801 - Brilliant green agar, modified

EMAIL THIS PAGE TO A FRIEND
B1801 Sigma

Brilliant green agar, modified

BioChemika, for microbiology

Purchase

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Solid Media/Agars, Media, Media by Application,
grade   for microbiology
product line   BioChemika
shelf life   limited shelf life, expiry date on the label
pH   6.9±0.2 (25 °C)
suitability   selective and differential for Salmonella

Description

Components

Components (g/L)
Proteose Peptone 10.00
Yeast Extract 3.00
Lactose 10.00
Sucrose 10.00
Sodium Chloride 5.00
Phenol Red 0.08
Brilliant Green 0.0125
Agar 20.00

Application

Used for selective isolation of Salmonellae other than Salmonella typhi.

Price and Availability


Sterility Testing
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?