• USA Home
  • B1802 - Brilliant green bile agar

EMAIL THIS PAGE TO A FRIEND
B1802 Sigma

Brilliant green bile agar

for microbiology

Purchase

Properties

Related Categories All Solid Media/Agars, Media, Microbiology More...
grade   for microbiology
pH   6.9±0.2 (25 °C)

Description

Components

Components (g/L)
Peptide Digest of Animal Tissue 8.25
Lactose 1.90
Sucrose Sulphite 0.205
Ferric Chloride 0.0295
Monopotassium Phosphate 0.0153
Erioglaucine 0.0649
Basic Fuchsin 0.0776
Ox Gall 0.00295
Brilliant Green 0.0000295
Agar 10.15

Application

Recommend for enumeration of the coliform bacteria in water and wastewater.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?