EMAIL THIS PAGE TO A FRIEND
B2011 Sigma

Boc-Asn(Xan)-OH

Synonym: Nα-Boc-Nγ-(9-xanthenyl)-L-asparagine, Nα-Boc-Nγ-xanthyl-L-asparagine

  • CAS Number 65420-40-8

  • Empirical Formula (Hill Notation) C22H24N2O6

  • Molecular Weight 412.44

  •  Beilstein Registry Number 5172403

  •  MDL number MFCD00066157

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?