• USA Home
  • B2052 - Baird Parker sulfa supplement

EMAIL THIS PAGE TO A FRIEND
B2052 Sigma

Baird Parker sulfa supplement

for microbiology

Purchase

Properties

Related Categories Media, Microbiology, Supplements More...
grade   for microbiology
packaging   vial of 50 mg sulphamethazine
storage temp.   2-8°C

Description

Application

Baird Parker Sulpha Supplement is an antibacterial supplement that can be used for the suppression of the Proteus species growing on Baird Parker Agar Base.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
WO9275000
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?