• USA Home
  • B2420 - Benzoic acid-carboxy-14C

EMAIL THIS PAGE TO A FRIEND
B2420 Sigma

Benzoic acid-carboxy-14C

acetone solution

Purchase

Properties

InChI Key   WPYMKLBDIGXBTP-WGGUOBTBSA-N
form   acetone solution
extent of labeling   1-20 mCi per mmol
mass shift   M+2
storage temp.   2-8°C

Description

Packaging

sealed ampule

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?