• USA Home
  • B2549 - Branched-chain amino acid transaminase bacterial

EMAIL THIS PAGE TO A FRIEND
B2549 Sigma

Branched-chain amino acid transaminase bacterial

recombinant, expressed in E. coli, lyophilized powder, ≥2 units/mg protein (biuret)

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of 2-ketoisovalerate to L-valine per min at pH 7.0 at 37 °C in the presence of L-glutamate and pyridoxal phosphate.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?