• USA Home
  • B2768 - p-Aminobenzamidine Sepharose® 6B

EMAIL THIS PAGE TO A FRIEND
B2768 Sigma

p-Aminobenzamidine Sepharose® 6B

aqueous ethanol suspension

Purchase

Properties

form   aqueous ethanol suspension
storage temp.   2-8°C

Description

Physical form

Suspension in 20% ethanol

Legal Information

Sepharose is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?