• USA Home
  • B2887 - 8-Bromoinosine 5′-diphosphate sodium salt

EMAIL THIS PAGE TO A FRIEND
B2887 Sigma

8-Bromoinosine 5′-diphosphate sodium salt

95-98%

  • CAS Number 102185-44-4

  • Empirical Formula (Hill Notation) C10H13BrN4O11P2

  • Molecular Weight 507.08

  •  MDL number MFCD00084711

  •  PubChem Substance ID 24891415

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotide Analogs More...
InChI Key   FTVMEMLAJADPSZ-UHFFFAOYSA-N
assay   95-98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?