• USA Home
  • B3027 - Sodium 2-methyl-2-propanethiolate

EMAIL THIS PAGE TO A FRIEND
B3027 Sigma

Sodium 2-methyl-2-propanethiolate

~97% (titration)

Synonym: 2-Methyl-2-propanethiol sodium salt, Sodium tert-butyl sulfide, t-Butanethiol sodium salt

  • CAS Number 29364-29-2

  • Linear Formula (CH3)3CSNa

  • Molecular Weight 112.17

  •  Beilstein Registry Number 3682306

  •  MDL number MFCD00043311

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   TUZFIWBNGKNFPW-UHFFFAOYSA-M
assay   ~97% (titration)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

71753 Sodium 2-methyl-2-propanethiolate, technical, ≥90% (RT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?