EMAIL THIS PAGE TO A FRIEND
B3032 Sigma

Boric oxide

~99% (titration)

Synonym: Boric anhydride, Boron oxide

Purchase

Properties

assay   ~99% (titration)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
ED7900000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?