• USA Home
  • B3305 - Bone Morphogenetic Protein Receptor IB/Fc Chimera human

EMAIL THIS PAGE TO A FRIEND
B3305 Sigma

Bone Morphogenetic Protein Receptor IB/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: BMPR-IB

Purchase

Properties

Related Categories BMPs and Receptors, Cell Biology, Cell Signaling and Neuroscience, Cytokines and Growth Factors, Cytokines, Growth Factors and Hormones,
recombinant   expressed in NSO cells
assay   >90% (SDS-PAGE)
form   lyophilized powder
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   human ... BMPR1B(658)

Description

Analysis Note

The biological activity is measured by its ability to inhibit recombinant human BMP-4-induced alkaline phosphatase production in C2C12 cells.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline

Biochem/physiol Actions

Bone Morphogenetic Protein Receptor IB/Fc Chimera, also known as activin receptor-like kinase (ALK)-6, is a type I serine/threonine kinase, which is required for the signal transduction of the TGF-β family cytokines. Human and murine bone morphogenetic protein receptor IB demonstrates 98% homology.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?