• USA Home
  • B3411 - (2R,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

EMAIL THIS PAGE TO A FRIEND
B3411 Sigma

(2R,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid

Synonym: Boc-(2R,3R)-3-amino-2-hydroxy-4-phenylbutyric acid

  • CAS Number 77171-41-6

  • Empirical Formula (Hill Notation) C15H21NO5

  • Molecular Weight 295.33

  •  Beilstein Registry Number 8993437

  •  MDL number MFCD00214254

Purchase

Properties

InChI Key   BHTRKISIDQZUQX-VXGBXAGGSA-N
storage temp.   −20°C

Description

Application

Novel protected amino acid for the preparation of epibestatin and analogs

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

15058 (2R,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?