• USA Home
  • B3430 - Bone Morphogenetic Protein Receptor II/Fc Chimera human

EMAIL THIS PAGE TO A FRIEND
B3430 Sigma

Bone Morphogenetic Protein Receptor II/Fc Chimera human

>97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: BMPR-II

Purchase

Description

Analysis Note

Recombinant human bone morphogenetic receptor II/Fc does not have BMP anatagonist activity. Its intended use is as a control for anti-BMPR-II.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Biochem/physiol Actions

BMP Receptor II/Fc is one of the five mammalian type II receptors for the TGF-β superfamily of ligands. It binds BMP-2, BMP-4, and BMP-7 weakly in the absence of type I receptor. Binding can be facilitated by the presence of BMP type I receptor. Human and murine bone morphogenetic protein receptor II demonstrates 97% homology.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?