• USA Home
  • B3514 - 2-(5-Bromo-2-pyridylazo)-5-(N-propyl-3-sulfopropylamino)aniline sodium salt

EMAIL THIS PAGE TO A FRIEND
B3514 Sigma

2-(5-Bromo-2-pyridylazo)-5-(N-propyl-3-sulfopropylamino)aniline sodium salt

≥90%

Synonym: 5-Br-PSAA

  • CAS Number 87035-60-7

  • Linear Formula C17H21BrN5O3SNa

  • Molecular Weight 478.34

Purchase

Properties

assay   ≥90%
mp   235-240 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?