• USA Home
  • B4006 - Bacillus subtilis (ATCC 6633)

EMAIL THIS PAGE TO A FRIEND
B4006 Sigma

Bacillus subtilis (ATCC 6633)

lyophilized cells

Purchase

Properties

Related Categories Biochemicals and Reagents, Cells, Microbiology, Microorganisms, Protein Extracts,
form   lyophilized cells
storage temp.   −20°C

Description

General description

Produced in pure culture - not intended for use as a starter culture. Not processed or packaged aseptically.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?