• USA Home
  • B4009 - Boc-Trp-Met-Asp-Phe amide

EMAIL THIS PAGE TO A FRIEND
B4009 Sigma

Boc-Trp-Met-Asp-Phe amide

≥97% (HPLC)

Synonym: Gastrin Related Tetrapeptide

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Cholecystokinins, Neuropeptides, Peptides and Proteins,
InChI Key   DDROKKYSSQPXQO-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?