• USA Home
  • B4292 - (S)-N-Benzyl-1-(1-naphthyl)ethylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
B4292 Sigma

(S)-N-Benzyl-1-(1-naphthyl)ethylamine hydrochloride

~98% (titration)

Synonym: (S)-N-[1-(1-Naphthyl)ethyl]benzylamine hydrochloride

  • CAS Number 163831-66-1

  • Empirical Formula (Hill Notation) C19H19N · HCl

  • Molecular Weight 297.82

  •  MDL number MFCD00214136

  •  PubChem Substance ID 24891764

Purchase

Properties

Related Categories Amine Salts, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   KCLJDAVYFCDIDY-RSAXXLAASA-N
assay   ~98% (titration)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?