• USA Home
  • B4503 - Plasma barium sulfate eluate from bovine plasma

EMAIL THIS PAGE TO A FRIEND
B4503 Sigma

Plasma barium sulfate eluate from bovine plasma

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Coagulation Proteins and Reagents, Plasma & Blood Proteins, Proteins and Derivatives
form   lyophilized powder
storage temp.   −20°C

Description

Other Notes

Contains prothrombin and factors VII, IX and X.

Unit Definition

One unit is equivalent to the eluate from one liter of plasma.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?