EMAIL THIS PAGE TO A FRIEND
B4505 Sigma

Behenyl acetate

~99%

Synonym: Acetic acid behenyl ester

  • CAS Number 822-26-4

  • Empirical Formula (Hill Notation) C24H48O2

  • Molecular Weight 368.64

  •  MDL number MFCD00056277

  •  PubChem Substance ID 24891779

Purchase

Properties

Related Categories Alcohol Acetates, Biochemicals and Reagents, Fatty Acyls, Fatty Esters, Lipids More...
InChI Key   AWYRDXDPCQRJHE-UHFFFAOYSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?