• USA Home
  • B4512 - N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine

EMAIL THIS PAGE TO A FRIEND
B4512 Sigma

N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine

~98%

Synonym: Bz-DMT-dC

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C30+, Chemical Synthesis, Heterocyclic Building Blocks,
InChI Key   MYSNCIZBPUPZMQ-VOTWKOMSSA-N
assay   ~98%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?