• USA Home
  • B4514 - N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine 3′-O-succinic acid

EMAIL THIS PAGE TO A FRIEND
B4514 Sigma

N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine 3′-O-succinic acid

~95%

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Inhibitors, Enzyme Inhibitors by Type, Enzymes, Inhibitors, and Substrates, Substrate Analogs More...
InChI Key   GJWDGMQWTLYOCO-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?