• USA Home
  • B4523 - Brilliant Blue G solution

EMAIL THIS PAGE TO A FRIEND
B4523 Sigma

Brilliant Blue G solution

0.8% (w/v) Brilliant Blue G in 12% (w/v) trichloroacetic acid

Purchase

Properties

InChI Key   RWVGQQGBQSJDQV-UHFFFAOYSA-M

Description

Analysis Note

This product has been tested for suitability on SDS-PAGE and IEF.

Application

Brilliant blue G – TCA will fix and stain protein in non-denaturing PAGE, SDS-PAGE and agarose gels (IEF) in 30-60 min with no destaining required. Fixing is not required prior to staining .

Other Notes

The pH has been adjusted with H2SO4 and KOH (0.8N and 0.9M final concentrations respectively) to remove insolubles.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1760 8/PG 2
WGK Germany 
2
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

B8522 Brilliant Blue G solution, Concentrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?