• USA Home
  • B4676 - Bordet Gengou Agar Base without Peptone

EMAIL THIS PAGE TO A FRIEND
B4676 Sigma

Bordet Gengou Agar Base without Peptone

BioChemika, for microbiology

Purchase

Properties

grade   for microbiology
product line   BioChemika
shelf life   limited shelf life, expiry date on the label
final pH   6.7±0.2 (25 °C)

Description

Components

Ingredients (g/L)
Potatoes infusion from, 125.00
Sodium chloride, 5.50
Agar, 20.00

Application

Recommended for the detection and isolation of Bordetella pertussis and Bordetella parapertussis.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?