• USA Home
  • B4682 - Octakis (6-O-t-butyldimethylsilyl)-γ-cyclodextrin

EMAIL THIS PAGE TO A FRIEND
B4682 Sigma

Octakis (6-O-t-butyldimethylsilyl)-γ-cyclodextrin

>90%

  • Empirical Formula (Hill Notation) C96H192O40Si8

  • Molecular Weight 2211.21

  •  MDL number MFCD03452845

  •  PubChem Substance ID 329773241

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Chelation/Complexation Compounds,
InChI Key   MKSDSPVSZRMRNY-YKBAZCJNSA-N
assay   >90%
storage temp.   2-8°C

Description

Application

Octakis (6-O-t-butyldimethylsilyl)-γ-cyclodextrin or DIACTGCD was used to determine the enantio-discrimination of mono-substituted styrene oxides.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?