• USA Home
  • B4764 - Tyr-Bradykinin acetate salt hydrate

EMAIL THIS PAGE TO A FRIEND
B4764 Sigma

Tyr-Bradykinin acetate salt hydrate

≥97% (HPLC)

  • Empirical Formula (Hill Notation) C59H82N16O13 · xC2H4O2 · yH2O

  • Molecular Weight 1223.38 (anhydrous free base basis)

  •  MDL number MFCD11045987

  •  PubChem Substance ID 24891701

Purchase

Properties

Related Categories Bradykinins, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   KBAUSVRYFSMFBT-PWPLERHXSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Tyr-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?