• USA Home
  • B4765 - Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Biotin antibody produced in sheep

EMAIL THIS PAGE TO A FRIEND
B4765 Sigma

Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Biotin antibody produced in sheep

affinity isolated antibody, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, MO-MR, Primary Antibodies
biological source   sheep
antibody form   affinity isolated antibody
clone   polyclonal
form   buffered aqueous solution
application(s)   direct ELISA: 1:20,000
conjugate   biotin conjugate
storage temp.   2-8°C

Description

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?