EMAIL THIS PAGE TO A FRIEND
B4804 Sigma
Acculab® VI Balance
VI-120 g - 1 mg
Purchase
Properties
Related Categories Acculab Balances & Accessories, Balances and Accessories, Labware More...
material   stainless steel platform
mfr. no.   VI-120 g - 1 mg
availability   not available in EU
measuring range   120 g, resolution: 0.001 g
overall size   115 mm × 133 mm × 235 mm
platform D   8 cm
Description
General description
All Acculab balances listed below
• tare over full capacity
• have keypad auto-calibration function
• have auto-off with memory to prolong battery life
• have four weighing modes (g, avoirdupois oz, troy oz, dwt), plus parts counting on V-400, V-600, V-1200, and V-2400 models
• LCD has overload/underload indicators and low battery indicator
• operate with fully compensated strain-gauge transducer technology, optimal between 18-25 °C
• AC adapter included
General-purpose laboratory balances with a wide range of capacities. Excellent value in precision weighing.
Legal Information
Acculab is a registered trademark of Globaltec Corp.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?