• USA Home
  • B4805 - Anti-BAFF antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
B4805 Sigma

Anti-BAFF antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-BLyS, Anti-TALL-1, Anti-THANK, Anti-TNFSF13B

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Death Receptor Ligands,
species reactivity   human, mouse, rat
application(s)   ELISA: suitable
  immunocytochemistry: suitable
  immunofluorescence: suitable
  western blot: suitable
clone   polyclonal
concentration   1 mg/mL
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 30 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... TNFSF13B(10673)
mouse ... Tnfsf13b(24099)
rat ... Tnfsf13b(498666)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the C-terminal amino acids 254-269 of human BAFF.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

PRS2221 Anti-BAFF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?