• USA Home
  • B5028 - Boc-γ-benzyl-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride

EMAIL THIS PAGE TO A FRIEND
B5028 Sigma

Boc-γ-benzyl-Glu-Gly-Arg-7-amido-4-methylcoumarin hydrochloride

~95%

  • CAS Number 133448-22-3

  • Empirical Formula (Hill Notation) C35H45N7O9 · HCl

  • Molecular Weight 744.23

  •  MDL number MFCD00133631

  •  PubChem Substance ID 24891805

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   GWPBOFRTQMAFMY-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?