• USA Home
  • B5055 - Monoclonal Anti-Phosphotyrosine–Biotin antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
B5055 Sigma

Monoclonal Anti-Phosphotyrosine–Biotin antibody produced in mouse

clone PY20, purified immunoglobulin, buffered aqueous glycerol solution

Synonym: Phospho-Tyr, Phospho-tyrosine, p-Tyr

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Phosphoproteins,
biological source   mouse
antibody form   purified immunoglobulin
clone   PY20, monoclonal
form   buffered aqueous glycerol solution
application(s)   western blot: 1:5,000
conjugate   biotin conjugate
shipped in   wet ice
storage temp.   −20°C

Description

Physical form

Solution in 20 mM phosphate buffered saline, pH 7.5, containing 50% glycerol and 3 mM sodium azide

General description

Reversible phosphorylation of proteins is an important post-translational modification that plays a regulatory role in the expression of most proteins in the cells. Reversible phosphorylation at multiple serine, tyrosine and threonine residues mediate numerous signalling pathways in both prokaryotic and Eukaryotic cells. Cellular proteins with phosphorylated tyrosine increase many fold by the activation of tyrosine kinases. Most mitogenic receptor systems such as EGF, PDGF, insulin receptors contain tyrosine kinase domains that undergo autophosphorylation when receptors bind to the respective ligands. Tyrosine-specific protein kinase activity has also been described in many retroviral oncogene proteins. Cells transformed by these oncogenes contain elevated levels of phosphotyrosine. Many of the oncogenes found in mammalian oncogenic viruses encode tyrosine protein kinases that reside in the cellular cytoplasm. Others encode transmembrane receptors whose tyrosine phosphotransferase activity is stimulated by the binding of ligand to the extracellular domain.
Monoclonal Anti-Phosphotyrosine is specific for both native and denatured proteins containing phosphorylated tyrosine. It does not react with either phosphoserine or phosphothreonine.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?