• USA Home
  • B5058 - Anti-Bone Morphogenetic Protein 1, N-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
B5058 Sigma

Anti-Bone Morphogenetic Protein 1, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym: Anti-BMP-1, Anti-ProCollagen C-Endopeptidase

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cytokines and Growth Factors, Antibodies to Extracellular Matrix,
species reactivity   human
application(s)   western blot (chemiluminescent): 1:5,000
  western blot: 1:1,000
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... BMP1(649)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the N-terminal end of human BMP-1 (ProCollagen C-Endopeptidase, PCP, mTld, EC 3.4.24.1)

Physical form

Solution in phosphate buffered saline, containing 50% glycerol and 0.01% sodium azide.

Specificity

Detects the shorter form of BMP-1, as well as the longer mTld form

Price and Availability


Antibody Explorer

Explore with Confidence

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?