• USA Home
  • B5151 - 4-(Boc-aminomethyl)phenyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
B5151 Sigma

4-(Boc-aminomethyl)phenyl isothiocyanate

~95%

Synonym: 4-(N-tert-Butoxycarbonylaminomethyl)phenyl isothiocyanate, tert-Butyl N-(4-isothiocyanatophenylmethyl)carbamate, BAMPITC, N-Boc-4-isothiocyanatobenzylamine

  • CAS Number 89631-74-3

  • Empirical Formula (Hill Notation) C13H16N2O2S

  • Molecular Weight 264.34

  •  Beilstein Registry Number 9557243

  •  MDL number MFCD00077918

Purchase

Properties

InChI Key   FZQIQTXXAATZOS-UHFFFAOYSA-N
assay   ~95%
storage temp.   2-8°C

Description

Application

Novel protein microsequencing reagent useful with more sensitive fluorogenic detection methods.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?