• USA Home
  • B5275 - (+)-Brompheniramine maleate salt

EMAIL THIS PAGE TO A FRIEND
B5275 Sigma

(+)-Brompheniramine maleate salt

Synonym: Dexbrompheniramine maleate salt

  • CAS Number 2391-03-9

  • Empirical Formula (Hill Notation) C16H19BrN2 · C4H4O4

  • Molecular Weight 435.31

  •  MDL number MFCD00072141

  •  PubChem Substance ID 24278271

Purchase

Properties

Related Categories Antagonists, Cell Biology, Cell Signaling and Neuroscience, Histaminergics, Neuroscience,
InChI Key   SRGKFVAASLQVBO-DASCVMRKSA-N
Gene Information   human ... HRH1(3269)

Description

Biochem/physiol Actions

Antihistamine; H1 histamine receptor antagonist.

Other Notes

Active isomer

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?