• USA Home
  • B5306 - [(2R,4R,5S)-2-Benzyl-5-(Boc-amino)-4-hydroxy-6-phenylhexanoyl]-Leu-Phe-NH2

EMAIL THIS PAGE TO A FRIEND
B5306 Sigma

[(2R,4R,5S)-2-Benzyl-5-(Boc-amino)-4-hydroxy-6-phenylhexanoyl]-Leu-Phe-NH2

>90% (TLC)

Synonym: L-685,458

  • Empirical Formula (Hill Notation) C39H52N4O6

  • Molecular Weight 672.85

  •  MDL number MFCD03452850

Purchase

Description

Application

Potent γ-secretase inhibitor containing a hydroxy­ethylene dipeptide isostere that should serve as a transition analogue to direct the inhibitor to the active site of an aspartyl protease target.

Price and Availability


Amplified Detection

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?