• USA Home
  • B5431 - Boc-Gln-D-Trp(Formyl)-Phe benzyl ester trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
B5431 Sigma

Boc-Gln-D-Trp(Formyl)-Phe benzyl ester trifluoroacetate salt

≥90% (HPLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
assay   ≥90% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Boc-Gln-D-Trp(For)-Phe-OBzl

Application

Antagonist of Substance P.

Biochem/physiol Actions

Boc-Gln-D-Trp(Formyl)-Phe benzyl ester was developed as a glutaminyl-D-tryptophylphenylalanine derivative Substance P antagonist.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?