EMAIL THIS PAGE TO A FRIEND
B5764 Sigma

N2-Benzoylguanosine

  • CAS Number 3676-72-0

  • Empirical Formula (Hill Notation) C17H17N5O6

  • Molecular Weight 387.35

  •  PubChem Substance ID 329773265

Purchase

Properties

InChI Key   WZKJANWNNBJHRS-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?