• USA Home
  • B5782 - Benzyl α-D-mannopyranoside

EMAIL THIS PAGE TO A FRIEND
B5782 Sigma

Benzyl α-D-mannopyranoside

≥98%

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Monosaccharide More...
InChI Key   GKHCBYYBLTXYEV-BNDIWNMDSA-N
assay   ≥98%
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?