• USA Home
  • B6154 - Brain natriuretic peptide-45 rat

EMAIL THIS PAGE TO A FRIEND
B6154 Sigma

Brain natriuretic peptide-45 rat

≥97% (HPLC)

Synonym: BNP-45

  • CAS Number 123337-89-3

  • Empirical Formula (Hill Notation) C213H349N71O65S3

  • Molecular Weight 5040.68

  •  MDL number MFCD00081416

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Natriuretic Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Ser-Gln-Asp-Ser-Ala-Phe-Arg-Ile-Gln-Glu-Arg-Leu-Arg-Asn-Ser-Lys-Met-Ala-His-Ser-Ser-Ser-Cys-Phe-Gly-Gln-Lys-Ile-Asp-Arg-Ile-Gly-Ala-Val-Ser-Arg-Leu-Gly-Cys-Asp-Gly-Leu-Arg-Leu-Phe [Disulfide Bridge: 23-39]

Other Notes

Storage form of BNP-32

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

B9901 Brain Natriuretic Peptide-32 rat, ≥97% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?