EMAIL THIS PAGE TO A FRIEND
B6501 Sigma

Boc-Gln-OH

Synonym: Nα-(tert-Butoxycarbonyl)-L-glutamine, Boc-L-glutamine

  • CAS Number 13726-85-7

  • Linear Formula NH2COCH2CH2CH(COOH)NHCOOC(CH3)3

  • Molecular Weight 246.26

  •  Beilstein Registry Number 2127805

  •  EC Number 237-296-8

  •  MDL number MFCD00065571

  •  PubChem Substance ID 329773278

Purchase

Properties

InChI Key   VVNYDCGZZSTUBC-LURJTMIESA-N
mp   113-116 °C (dec.)(lit.)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?