• USA Home
  • B6519 - 6-Bromo-2-naphthyl β-D-glucuronide

EMAIL THIS PAGE TO A FRIEND
B6519 Sigma

6-Bromo-2-naphthyl β-D-glucuronide

Synonym: 6-Bromo-2-naphthyl β-D-glucopyranosiduronic acid

Purchase

Description

Substrates

A histochemical substrate for β-D-glucuronidase.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?