• USA Home
  • B6780 - Bio-11-dUTP tetralithium salt

EMAIL THIS PAGE TO A FRIEND
B6780 Sigma

Bio-11-dUTP tetralithium salt

For DNA labeling

Synonym: 5-(N-[N-Biotinyl-ε-aminocaproyl]-3-aminoallyl)-2′-deoxyuridine 5′-triphosphate, Bio-11-dUTP

  • CAS Number 120964-50-3

  • Empirical Formula (Hill Notation) C28H45N6O17P3S

  • Molecular Weight 862.67

Purchase

Properties

grade   for molecular biology
form   powder
shipped in   dry ice
storage temp.   −20°C

Description

Quantity

Vial contains 50 nanomoles

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?