EMAIL THIS PAGE TO A FRIEND
B6806 Sigma

Bomin-3

≥80% (TLC), solid

Synonym: 2-Butoxy-2-oxo-3H-1,4,2-benzodioxaphosphorine

  • Empirical Formula (Hill Notation) C11H15O4P

  • Molecular Weight 242.21

  •  MDL number MFCD00285577

  •  PubChem Substance ID 329773284

Purchase

Properties

Related Categories A to C, Biochemicals and Reagents, Carboxylesterase, Enzyme Inhibitors, Enzyme Inhibitors by Enzyme,
InChI Key   SPJHCIRTIZORLB-UHFFFAOYSA-N
assay   ≥80% (TLC)
form   solid

Description

Biochem/physiol Actions

Selective irreversible inhibitor of carboxylesterases.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?