EMAIL THIS PAGE TO A FRIEND
B6885 Sigma

Biotin−Agarose

saline suspension

Synonym: Vitamin H Agarose

Purchase

Properties

Related Categories Affinity Chromatography, Avidin Biotin Matrices, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
extent of labeling   2-6 μmol per mL
matrix activation   epoxy
matrix attachment   carboxy (amide linkage through diaminopropane spacer); resin contains residual amino groups which are charged at physiological pH
matrix spacer   17 atoms
capacity   20-30 mg/mL binding capacity (avidin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.01% thimerosal

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?