• USA Home
  • B7031 - S-(N-Benzylthiocarbamoyl)-L-cysteine

EMAIL THIS PAGE TO A FRIEND
B7031 Sigma

S-(N-Benzylthiocarbamoyl)-L-cysteine

  • CAS Number 35446-36-7

  • Empirical Formula (Hill Notation) C11H14N2O2S2

  • Molecular Weight 270.37

  •  MDL number MFCD00133804

  •  PubChem Substance ID 24891968

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   RSTSYGXUDMJEFP-UHFFFAOYSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Glutathione S-transferase inducer

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?