• USA Home
  • B7530 - α-Boc-Deacetylleupeptin

EMAIL THIS PAGE TO A FRIEND
B7530 Sigma

α-Boc-Deacetylleupeptin

≥97% (TLC)

Synonym: Nα-t-Boc-Leu-Leu-Arg-al

  • CAS Number 81344-47-0

  • Empirical Formula (Hill Notation) C23H44N6O5

  • Molecular Weight 484.63

  •  MDL number MFCD00133579

  •  PubChem Substance ID 24892002

Purchase

Description

Biochem/physiol Actions

Inhibits fertilization by preventing penetration of the zona pellucida by sperm cells.

Price and Availability


Amplified Detection

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?