• USA Home
  • B7804 - Anti-Bonzo antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
B7804 Sigma

Anti-Bonzo antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-STRL33, Anti-TYMSTR

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Aids and Viral Research, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
species reactivity   human
application(s)   western blot: 0.5 μg/mL using human SW1353 cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CXCR6(10663)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 319-338 of the C-terminal of human Bonzo (STRL33, TYMSTR). The sequence is identical in macaque and African green monkey.

Physical form

Solution in phosphate buffered saline, containing 0.02% sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?